ContactUsLinkComponent
Eyewear
Uvex Genesis® Protective Eyewear, Honeywell Safety
# de catalogue 10790-624
Eyewear
Uvex Genesis® Protective Eyewear, Honeywell Safety
# de catalogue 10790-624
Spécifications
  • Lens Coating:
    Anti-fog (HydroShield™)
  • Lens Tint:
    Clear
  • Frame Color:
    Black
  • Supplier No.:
    S3200HS
Spécifications
A propos de cet article
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance.
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance. The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility. Soft, flexible “fingers” adjust and conform to any nasal profile, minimizing slippage. A three-position ratchet lens inclination system completes the custom fit. A ventilation channel between frame and lens vents the eye socket area for outstanding moisture/fog control. Lens replacement is quick, easy, and economical. Ultra-dura® lens coating provides superior scratch and chemical resistance. Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme® lens coating features Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Clear lenses are appropriate for most indoor applications. Gold mirror, amber, and espresso lenses minimize the effects of sunlight and glare. The SCT-Reflect 50 lenses are designed for use in work applications with varying light conditions. This tint is a comfortable solution for workers who move between shaded or indoor locations and direct sunlight. SCT-Vermillion lenses are ideal for indoor applications requiring contrast, such as inspection. SCT-Blue lenses help counteract sodium vapor lighting. The Patriot® RWB frame features a red, white, and blue design with miniature U.S. flags at temples and “United We Stand” inscribed on bottom right lens outside field of vision.
Eyewear meets military Vo ballistic test for impact and ANSI Z87+ and CSA Z94.3 standards.
  • Dual 9-base wraparound lens provides uncompromised peripheral vision and protection Elastomer brow-guard diffuses and deflects impact
  • Temple length and lens inclination are adjustable
  • Soft fingers conform to any nasal profile
  • Frames guaranteed for life
The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility.
Clear lenses are appropriate for most indoor applications. Gold mirror, amber, and espresso lenses minimize the effects of sunlight and glare. The SCT-Reflect 50 lenses are designed for use in work applications with varying light conditions. This tint is a comfortable solution for workers who move between shaded or indoor locations and direct sunlight. SCT-Vermillion lenses are ideal for indoor applications requiring contrast, such as inspection. SCT-Blue lenses help counteract sodium vapor lighting. The Patriot® RWB frame features a red, white, and blue design with miniature U.S. flags at temples and 'United We Stand' inscribed on bottom right lens outside field of vision.
Soft, flexible 'fingers' adjust and conform to any nasal profile, minimizing slippage. A three-position ratchet lens inclination system completes the custom fit. A ventilation channel between frame and lens vents the eye socket area for outstanding moisture/fog control. Lens replacement is quick, easy, and economical. Ultra-dura® lens coating provides superior scratch and chemical resistance. Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme® lens coating features Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Certifications: Eyewear meets military Vo ballistic test for impact and ANSI Z87+ and CSA Z94.3 standards.
Customers Who Bought This Also Bought