ContactUsLinkComponent
Solid Steel Ball
Smashing Steel Sphere Kinetic Energy Demo
# de catalogue 470163-918
Fournisseur:  American Scientific
Solid Steel Ball
Smashing Steel Sphere Kinetic Energy Demo
# de catalogue 470163-918
Fournisseur:  American Scientific
Spécifications
  • Description:
    Smashing Steel Sphere Kinetic Energy Demo
  • Cat. no.:
    470163-918
  • Supplier No.:
    3484-38
Spécifications
A propos de cet article
For physics experiments and demonstrations such as kinetic, density and volume measurements, pendulum bob, potential energy and conservation experiments, ballistics etc. Solid ball, 1-1/2" (40mm) diameter. Made of Steel. Sold as a pair. Comes with a Teacher's Guide.
Customers Who Bought This Also Bought