Order Entry
ContactUsLinkComponent
Recombinant Murine IFN-Lambda2
Mouse Recombinant IFN-λ2 (from E. coli)
# de catalogue 10773-396
Fournisseur:  Peprotech
Numéro CAS:  
Mouse Recombinant IFN-λ2 (from E. coli)
# de catalogue 10773-396
Fournisseur:  Peprotech
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Gene ID:
    330496 Q4VK74
  • Protease-free:
    Y
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Conjugation:
    Unconjugated
  • Size:
    250 μg
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protein synonyms:
    IL-28A
  • Protein/peptide name:
    IFN-lambda 2
  • Purity:
    0.98
  • Molecular weight:
    19.8 kDa
  • Sequence:
    MDPVPRATRL PVEAKDCHIA QFKSLSPKEL QAFKKAKDAI EKRLLEKDMR CSSHLISRAW DLKQLQVQER PKALQAEVAL TLKVWENMTD SALATILGQP LHTLSHIHSQ LQTCTQLQAT AEPKPPSRRL SRWLHRLQEA QSKETPGCLE DSVTSNLFRL LTRDLKCVAS GDQCV
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient
  • Cat. no.:
    10773-396
  • Origin:
    USA
  • Supplier No.:
    250-33-250UG

Spécifications

A propos de cet article

IFN λ1, 2, and 3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and interferons. All three IFN-lambdas use a distinct receptor system composed of an IFN- λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defenses similar to, but distinct from, that of type I interferons. Recombinant Murine IFN- λ2 is a 19.8 kDa protein containing 175 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Customers Who Bought This Also Bought