Innovating Science® Dialysis Tubings
Catalog # 470163-406
Supplier: Ward's Science
Specifications
- Ø:29.6 mm
- Width:45 mm
- Description:Dialysis tubing
- Length:30 m
- Cat. no.:470163-406
- Supplier No.:ADDD0107-100
Specifications
About this item
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
- Available in multiple lengths
Flat width and open diameter measurement provided in the ordering table.
Customers Who Bought This Also Bought