Innovating Science® Dialysis Tubings
# de catalogue 470163-402
Fournisseur: Ward's Science
Spécifications
- Ø:15.9 mm
- Length:30 m
- Width:25 mm
- Description:Dialysis tubing
- Supplier No.:ADDD0106-100
Spécifications
A propos de cet article
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
- Available in multiple lengths
Flat width and open diameter measurement provided in the ordering table.
Customers Who Bought This Also Bought