Reagent Reservoirs, VistaLab
Catalogue # 10770-300
Fournisseur: CELLTREAT Scientific Products
Spécifications
- Sterility:Non sterile
- Capacity:10 ml
- Description:One-Compartment Tray; Mini Reservoir
- Packed:Bulk
- Cat. no.:10770-300
- Supplier No.:4054-1012
Spécifications
À propos de cet article
VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
- Trough-within-a-trough design for greatest sample recovery
- Pour-off spouts at every corner
- Wider base for stability
- Free of animal products
- Sterile reservoirs are RNase/DNase-free and non-pyrogenic
- Bulk or individually wrapped
- 12-channel polypropylene reservoir is autoclavable
Sterile and non-sterile reservoirs and reservoir coolers are available for purchase, with bulk or individually wrapped formats for compatibility with any distribution method.
Frequently Bought Together