Order Entry
ContactUsLinkComponent
Jug
Safety Coated Glass Jugs, Amber
Catalog # 10770-320
Supplier:  Qorpak
Product available on GSA Advantage®
Jug
Safety Coated Glass Jugs, Amber
Catalog # 10770-320
Supplier:  Qorpak
Product available on GSA Advantage®

Specifications

  • Cap size:
    38-430
  • Cap Type:
    Black phenolic F217 and fluoropolymer resin liner
  • Capacity:
    4 L (135 oz.)
  • Description:
    Safety coated amber jug with pour out neck finish, cap attached
  • Cat. no.:
    10770-320
  • Treatment:
  • Supplier No.:
    259410

Specifications

About this item

Safety coated amber jugs with molded finger grips are the perfect storage solution for light sensitive products.

  • Amber glass protects against harmful UV rays
  • Jugs are slip and impact resistant, making them ideal for handling volatile substances
  • Choose jugs only, or jugs with attached Black Phenolic F217 and PTFE lined caps
Customers Who Bought This Also Bought