Acid, Caustic and Solvent Spill Kit
# de catalogue 470163-134
Fournisseur: Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Caustic Neutralizer, 2 kg
- Supplier No.:AD-SS0917-2KG
Spécifications
A propos de cet article
Be prepared for spills in your lab.
- Personal Protective Equipment
- Diatomaceous Earth
- Vermiculite
- Dustpan and Brush Set
- Storage Container for Safe Disposal
Splash-resistant goggles protect while kits for caustic and acid spills clean up to one liter of four solvents.
A storage container allows for safe disposal after cleaning.
Ordering information: This kit contains enough material to clean up a liter of four solvents in the lab. It is complete with instruction sheets and SDS. The kit contains diatomaceous earth, an acid spill clean-up kit, a caustic clean-up kit, vermiculite, splash-resistant goggles, nitrile gloves, a dustpan and brush set, and a spill kit storage container. Replacement materials are available separately for you to simply restock your kit after a spill.
Customers Who Bought This Also Bought