ContactUsLinkComponent
Electrochemical Series
Innovating Science® The Electrochemical Series
# de catalogue 470163-124
Fournisseur:  Ward's Science
Electrochemical Series
Innovating Science® The Electrochemical Series
# de catalogue 470163-124
Fournisseur:  Ward's Science
Spécifications
  • Description:
    The electrochemical series, full kit
  • Cat. no.:
    470163-124
  • Supplier No.:
    IS8030
  • No. of Uses:
    15
Spécifications
A propos de cet article
What Are Redox Potentials?
  • Classic AP Chemistry Experiment
  • Instructions Included
Students will study electrochemical series. The electrochemical series is built up by arranging various redox equilibria in order of their standard electrode potentials (redox potentials). When a strip of metal (an electrode) is placed in water the metal has a tendency to go into solution as ions, with a simultaneous build up of electrons on the metal strip. This process produces an electrical potential difference between the metal and solution which is called an electrode potential (Eº).
Ordering information: Includes Copper Metal Strips, Zinc Metal Strips, Magnesium Metal Strips, Iron Metal Strips, Aluminum Metal Strips, Filter Paper Strips, Copper Sulfate EZ-Prep, Zinc Sulfate EZ-Prep, Magnesium Sulfate EZ-Prep, Iron Sulfate EZ-Prep, Aluminum Sulfate EZ-Prep, Teacher's Guide and Student Copymasters.
Delivery: Instructor's manual and SDS included.
Caution: Classic AP Chemistry Experiment Instructions Included
Customers Who Bought This Also Bought