HEPA filter, Labconco
Catalog # 10769-428
Supplier: Labconco
Specifications
- Depth:30"
- Width:16"
- Description:HEPA Filter
- Height:4"
- Cat. no.:10769-428
- Weight:11 lbs.
- Supplier No.:9577401
Specifications
About this item
The optional HEPA Filter, 99.995% efficient, H14 type, for non-hazardous particulate capture.
Total HEPA Filter weight per model: 3' Hood - 10 lbs; 4' Hood - 20 lbs; 5' Hood - 30 lbs; 6' Hood - 40 lbs; 8' Hood - 50 lbs.
Ordering information: For use with Protector® Filtered Fume Hoods. Order quantity required per model; 3' Hood - 1 filter; 4' Hood - 2 filters; 5' Hood - 3 filters; 6' Hood - 4 filters; 8' Hood - 5 filters.
Customers Who Bought This Also Bought