Silica sand 30 Mesh
# de catalogue 80705-016
Fournisseur: Avantor
Numéro CAS: 112945-52-5
Spécifications
- Packaging:Bag
- Size:22.7 kg
- Cat. no.:80705-016
- Supplier No.:AMAS029-944-01
Spécifications
A propos de cet article
:Silica
Point d'ébullition:2230 °C (1013 hPa)
Point de Fusion:1427 °C
Densité:2.622 to 2.645 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:30 Mesh
:Boiling Pt:
:Boiling Pt: 2230 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 1427 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.083 g/mol
Température d'entreposage:Ambient
EINECS:238-878-4
:GHS08
Customers Who Bought This Also Bought