Filter Discs, Screen Support, Ace Glass Incorporated
# de catalogue 10767-246
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:Fabric Screen Material (by the Yard), 350 micron
- Cat. no.:10767-246
- Supplier No.:5814-104
Spécifications
A propos de cet article
These filter discs have a wide variety of uses.
Available in highly absorbent paper, polypropylene, nylon, or PTFE.
Ordering information: For filter discs made from polyethylene, fluorocarbon, or other materials, please contact your Avantor representative.
Customers Who Bought This Also Bought