Spécifications
- Cap Size:N/A
- Capacity:-
- Sterility:Non sterile
- Description:Stopcock for VWR® Carboys
- Material:N/A
- Neck I.D.:—
Spécifications
A propos de cet article
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
- Non sterile
- Polypropylene and HDPE formats available
- Clear graduation marks
- Excellent chemical resistance
- Wide mouth
- Heavy duty stainless steel handle for easy carrying and pouring
Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak proof under standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
The high-density polyethylene, disposable carboys are ideal for storage and transfer of sterile fluids and pharmaceutical/biotech reagents. Carboys offer excellent chemical resistance and are well suited for use from –100 to 100 °C (–148 to 212 °F). Graduated to identify the volume despensed.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.