Order Entry
ContactUsLinkComponent
VWR* Carboy 20l rectangular spigot - high density polypropylene
VWR® Rectangular Carboy with Spigot, Wide Mouth
Catalog # 10755-106
Supplier:  VWR International
VWR* Carboy 20l rectangular spigot - high density polypropylene
VWR® Rectangular Carboy with Spigot, Wide Mouth
Catalog # 10755-106
Supplier:  VWR International

Specifications

  • Cap size:
    100 mm
  • Sterility:
    Non sterile
  • Capacity:
    20 L (5.3 gal.)
  • Description:
    Carboy 20L Rectangular Spigot - HDPE
  • Material:
    HDPE
  • Cat. no.:
    10755-106
  • Neck I.D.:
    85 mm

Specifications

About this item

Use for storing and dispensing solutions and media. Ideal for sterile water storage.

  • Non sterile
  • Polypropylene and HDPE formats available
  • Clear graduation marks
  • Excellent chemical resistance
  • Wide mouth
  • Heavy duty stainless steel handle for easy carrying and pouring

Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak proof under standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.

The high-density polyethylene, disposable carboys are ideal for storage and transfer of sterile fluids and pharmaceutical/biotech reagents. Carboys offer excellent chemical resistance and are well suited for use from –100 to 100 °C (–148 to 212 °F). Graduated to identify the volume despensed.

Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.

Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Ordering information: Spigot included. Always disassemble spigot and cap from carboy before autoclaving.
Customers Who Bought This Also Bought