BRAND PCR 8- and 12-Strip Tubes and Caps, BrandTech®
# de catalogue 10755-234
Fournisseur: Brandtech
Spécifications
- Cap Type:Flat (qPCR)
- Capacity:—
- Format:Strip
- Color:Clear
- Description:8-Cap Strips for PCR Plates
- Cat. no.:10755-234
- Supplier No.:781413
Spécifications
A propos de cet article
BRAND PCR tube strips and strip caps are manufactured from high quality virgin resins without lubricants or chemical additives for reproducible results in PCR applications.
- Tubes are autoclavable
- Supplied RNase-, DNase- and DNA-free
New Rigid strips with attached caps for easy handling. Flexible strips for easy strip division.
Strip tubes without attached caps have a tab on the end for easy handling.
Customers Who Bought This Also Bought