VWR® Lens Cleaning Tissues
Catalog # 10754-964
Supplier: VWR International
Specifications
- Dimensions:12.7 x 17.8 cm (5 x 7")
- Cat. no.:10754-964
- Packaging:2000 Sheets/Box
Specifications
About this item
Dust-free lens tissues are ideal for use with liquid cleaners.
- High wet strength
Clean safety glasses and goggles, microscope lenses, and more with no scratching.
Customers Who Bought This Also Bought