Order Entry
ContactUsLinkComponent
Microscope Cover Glass
Cover Glasses, Globe Scientific
# de catalogue 10118-661
Fournisseur:  Globe Scientific
Microscope Cover Glass
Cover Glasses, Globe Scientific
# de catalogue 10118-661
Fournisseur:  Globe Scientific

Spécifications

  • Numbering:
    #1
  • Description:
    Cover glasses, rectangular
  • L×W:
    40×22 mm
  • Cat. no.:
    10118-661
  • Supplier No.:
    1411-10

Spécifications

A propos de cet article

Globe Scientific Cover Glasses are packaged 1 ounce per pack, 10 packs/box (10 oz.) Each flip-top pack contains a desiccant and is wrapped in aluminum to prevent sticking.

Cover Glasses are available in three thicknesses: No 1 (0.14mm), No 1.5 (0.175mm), and No 2 (0.21mm).

Cover Glasses are available in three thicknesses: No 1 (0.14mm), No 1.5 (0.175mm), and No 2 (0.21mm).

Customers Who Bought This Also Bought