Orthophosphoric acid ≥85%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH153155E
Fournisseur: VWR International
Numéro CAS: 7664-38-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:500 ml
- Cat. no.:BDH153155E
- Supplier No.:BDH153155E
Spécifications
Résultats des tests de spécification
- Assay:Min. 85 %
- Volatile acids:Max. 0.0002 mEq/g
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 2 ppm
- Transmittance (254 nm) (0.1 mol/l):Min. 80 %
- Orthophosphoric acid ≥85%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Phosphoric acid
Point de Fusion:28 °C
Densité:1,71 to 1.87 g/cm³ (25 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: ~ 158 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: ~ 21 °C
:Pt Eclair:|Point eclaire:
:M.W. 98 g/mol
Température d'entreposage:Température ambiante
EINECS:231-633-2
:21
:158
ONU:1805
:GHS05
:12,07500
:14,07344
:Storage Temperature: Ambient
Densité:1.71 to 1.87 g/cm³ (25 °C)
:85
:Boiling Pt:
:Boiling Pt: ~ 158 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 28 °C
:Flash Pt:
:Flash Pt:
:8
:III
Température d'entreposage:Ambient
Customers Who Bought This Also Bought