1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH152803U
Fournisseur: VWR International
Numéro CAS: 5324-84-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:HDPE bottle
- Size:100 g
- Supplier No.:BDH152803U
Spécifications
Résultats des tests de spécification
- Assay:Min. 99 %
- IR Spectrum:Passes test
- pH (10 %):5.5-7.5
- Loss on drying (120°C; under vacuum):Max. 2.0 %
- Transmittance (200 nm) (0.005 mol/l):Min. 70 %
- Transmittance (220 nm) (0.005 mol/l):Min. 90 %
- Transmittance (250 nm) (0.005 mol/l):Min. 96 %
- 1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sodium octane-1-sulphonate
Point de Fusion:? 300 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: Min. 300 °C
:Pt Eclair:|Point eclaire:
:M.W. 216,28 g/mol
:Acide 1-octanesulfonique sel de sodium
Température d'entreposage:Ambient
EINECS:226-195-4
:GHS07
Point de Fusion:> 300 °C
Température d'entreposage:Température ambiante
:99
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: > 300 °C
:Flash Pt:
:Flash Pt:
:M.W. 216.28 g/mol
:1-Octanesulphonic acid sodium salt
Customers Who Bought This Also Bought