Lovibond MD 100 Colorimeter, Copper Range: 0.05-5.0 mg/L (Powder Reagents), Reagent included
# de catalogue 10752-382
Fournisseur: Lovibond
Lovibond MD 100 Colorimeter, Copper Range: 0.05-5.0 mg/L (Powder Reagents), Reagent included
# de catalogue 10752-382
Fournisseur: Lovibond
Customers Who Bought This Also Bought
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Fournisseur: Ricca Chemical
$205.73
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Fournisseur: MilliporeSigma
$1,352.49
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$795.94 to $2,497.03