Spécifications
- Volume:200 µl (1-200 µl)
- Environmentally Preferable:Waste Reducing
- Tip Color:Natural
- Packed:960 Tips/Pack
- Sterile:+
- Tip Style:Round tip
- Supplier No.:4144
Spécifications
A propos de cet article
These tip stations maximize bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
- Ideal single and mult-ichannel pipetting stations
- Five-layer deck design (2×96) matches the microtiter plate format
- Stable pipetting platform fits easily into sterile hood environments
- Lids can be snap-positioned to expose a single side (96 tips) only
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities. These tip stations are offered both sterile and non sterile.
Certified Free of: RNase, DNase, Pyrogens, PCR Inhibitors.
Both the hinged racks and relaod decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tip s are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.