Spécifications
- Description:Optical lens cleaner
- Size:60 ml (2 oz.)
- Packaging:6 lenses/pack, 8 packs/case
- Supplier No.:1100
Spécifications
A propos de cet article
Optical lens cleaner is a gentle cleaning solution that can be used to clean all optical glass surfaces including microscope optics, absorption cells, and lenses.
This cleaner contains no abrasives or silicones so it will not etch, scratch, or streak.