ContactUsLinkComponent
Flask
VWR® Volumetric Flask with PE Stopper, Class A
Catalog # 10545-984
Supplier:  VWR International
UL Listed
Flask
VWR® Volumetric Flask with PE Stopper, Class A
Catalog # 10545-984
Supplier:  VWR International
UL Listed
Specifications
  • Capacity:
    5 ml
  • Cat. no.:
    10545-984
  • Stopper Type:
    Polyethylene
  • Stopper Size:
    9
  • Certification:
    Batch
Specifications
About this item
These volumetric flasks are batch certified and are calibrated at 20 degrees Celsius To Contain.
  • Borosilicate glass fabrication
  • Polyethylene stopper construction
  • Durable white marking spot
They feature a single graduation ring and polyethylene stopper with marking spot.
Made from ASTM E438 Type 1 Class A Borosilicate 33 expansion glass as per ASTM specification E237 and E288. Durable white color printing which has good life in laboratory environment.
Customers Who Bought This Also Bought