Order Entry
Canada
ContactUsLinkComponent
VWR® Test Tubes with Sturdy Lip, Borosilicate Glass
Catalogue # 10545-928
Fournisseur:  VWR International
UL Listed
undefined
VWR® Test Tubes with Sturdy Lip, Borosilicate Glass
Catalogue # 10545-928
Fournisseur:  VWR International
UL Listed

Spécifications

  • Dimensions:
    25 x 150 mm
  • Cat. no.:
    10545-928

Spécifications

À propos de cet article

High quality test tubes designed and constructed for optimal performance.

  • High quality borosilicate glass construction
  • Durable white ceramic enamel marking spot

Allows use at temprature up to 500 °C and have excellent resistance to chemcial attack.

Tubes feature a sturdy lip and uniform wall thickness for maximum heat transfer and chemical resistance.

Certifications: Made from ASTM 438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E982.
Frequently Bought Together