Order Entry
ContactUsLinkComponent
Erlenmayer flask
VWR® Erlenmeyer Flasks, Narrow Mouth
Catalog # 10545-844
Supplier:  VWR International
UL Listed
undefined
VWR® Erlenmeyer Flasks, Narrow Mouth
Catalog # 10545-844
Supplier:  VWR International
UL Listed

Specifications

  • Graduation Interval:
    100 ml
  • Graduation Range:
    600 - 2000 ml
  • Capacity:
    2000 ml
  • Stopper No.:
    10

Specifications

About this item

Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.

  • Borosilicate glass fabrication
  • Durable design

Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.

Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.

Frequently Bought Together