Order Entry
ContactUsLinkComponent
Erlenmayer flask
VWR® Erlenmeyer Flasks, Narrow Mouth
# de catalogue 10536-926
Fournisseur:  VWR International
UL Listed
Erlenmayer flask
VWR® Erlenmeyer Flasks, Narrow Mouth
# de catalogue 10536-926
Fournisseur:  VWR International
UL Listed

Spécifications

  • Capacity:
    500 ml
  • Graduation Interval:
    50 ml
  • Graduation range:
    100 - 500 ml
  • Stopper No.:
    7

Spécifications

A propos de cet article

Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.

  • Borosilicate glass fabrication
  • Durable design

Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.

Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.

Customers Who Bought This Also Bought