VWR® Flat-Bottom Boiling Flasks
# de catalogue 10536-648
Fournisseur: VWR International
UL Listed
Spécifications
- Capacity:500 ml
- Neck Size:24/40
Spécifications
A propos de cet article
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
- Borosilicate glass fabrication
- Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Customers Who Bought This Also Bought