Specifications
- Temperature range:32 to 131ºF and 0 to 55ºC
- Description:Digital Barometer
- Humidity range:5 to 95% RH
- Cat. no.:10510-922
- Pressure Range:500 - 1030 (mbar)
Specifications
About this item
Perfect unit for plants, labs, and all other environments where there is a need to monitor conditions of environmental air parameters.
A four-line LCD continously and simultaneously shows barometric presure, temperature, relative humidity, barometric pressure trend and time.
Computer output allows unit to be connected to a computer using the accessory Computer Data Acquisition System (sold separately) to permanently capture readings.
Minimum/Maximum readings for barometric pressure, temperature and relative humidity are stored every hour for last 24 hours and are available for recall at the touch of a button. Barometric trend shows current reading, previous 1, 3, 6,12 hour and 24-hour readings in bar chart format; Adjustable altitude compensation from -100 to 2500 meters.