Tubing, Sulfur ready stainless steel, 1/8inch OD(3.175mm) x 0.085inch ID(2.159 mm), GC Grade 50ft
# de catalogue 10510-902
Fournisseur: General Separation Technologies, Inc.
Tubing, Sulfur ready stainless steel, 1/8inch OD(3.175mm) x 0.085inch ID(2.159 mm), GC Grade 50ft
# de catalogue 10510-902
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$72.06 to $624.50
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.66 to $576.72
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$291.56
Preassembled Vial Kits, 8-425, Screw-Thread, Unmarked Vials, Septa and Caps, Restek
Fournisseur: Restek
$764.77
SureSTART™ Total Recovery Glass Snap Top Microvials, 1.5 ml for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$342.01 to $346.57
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$180.74
SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
$547.20 to $567.60