Stainless Steel Packed Column: 6FT X 1/8 OD X 2MM ID, Molecular sieve 13X 80/100
# de catalogue 10499-730
Fournisseur: General Separation Technologies, Inc.
Stainless Steel Packed Column: 6FT X 1/8 OD X 2MM ID, Molecular sieve 13X 80/100
# de catalogue 10499-730
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$123.78
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$224.41
SureSTART™ MSCERT Screw Vial and Cap Kits, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$144.16 to $151.40
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$204.47
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$205.86
SureSTART™ High Recovery Glass Snap Top Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$468.01 to $511.40
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$112.24