Water, Molecular Biology Grade
# de catalogue VWRL0201-1000
Fournisseur: VWR
Product available on GSA Advantage®
Numéro CAS: 7732-18-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:PETG bottle, clear
- Size:1 L
- Supplier No.:02-0201-1000
Spécifications
Résultats des tests de spécification
- pH:check and record
- Sterility:negative
- Total Organic Carbon (before packaging):Max. 500 ppb
- Conductivity (before packaging):Max. 1.3 µs/cm
- Endotoxin:Max. 0.25 EU/mL
- DNAse:none detected
- RNAse:none detected
- Protease:none detected
- Water, Molecular Biology Grade:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydrogen oxide
Formule:H₂O
Point d'ébullition:100 °C (1013 hPa)
Point de Fusion:0 °C
Densité:1 g/cm³ (4 °C)
Numéro MDL:MFCD00011332
Numéro CAS:7732-18-5
:Eau
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:Filtered through a 0.2 µm filter.
:Point d'ébullition:
:Pt ébul.: 100 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: 0 °C
:Point eclaire:
:Flash Pt:
:M.W. 18,02 g/mol
Index Merck:13,10098
EINECS:231-791-2
:100 °C (1013 hPa)
Température d'entreposage:Température ambiante
:Filter 0,2 um
:12,10175
Customers Who Bought This Also Bought