n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH83632.400
Fournisseur: VWR International
Numéro CAS: 109-66-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:4 L
- Cat. no.:BDH83632.400
- Supplier No.:BDH83632.400
Spécifications
Résultats des tests de spécification
- Assay (by GC-FID) (as n-Pentane):Min. 99 %
- Water (by KF coulometry):Max. 0.01 %
- Nonvolatile Residue:Max. 0.0005 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Fluorescence (at emmission maximum for impurites) (254 nm):Max. 0.001 ppm
- Fluorescence (at emmission maximum for impurites) (365 nm):Max. 0.001 ppm
- Transmittance (200 nm):Min. 10 %
- Transmittance (210 nm):Min. 40 %
- Transmittance (215 nm):Min. 70 %
- Transmittance (222 nm):Min. 90 %
- Transmittance (240 nm):Min. 98 %
- n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Pentane
Formule:H₃C(CH₂)₃CH₃
Point d'ébullition:35 - 36 °C (1013 hPa)
Point de Fusion:-130 °C
Densité:0,63 g/cm³ (20 °C)
Point d'éclair:-40 °C
Numéro MDL:MFCD00009498
Numéro CAS:109-66-0
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 35 à 36 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -130 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: -40 °C
:M.W. 72,15 g/mol
Index Merck:13,07193
:n-Pentane
Point d'ébullition:35…36 °C (1013 hPa)
EINECS:203-692-4
:-130
Point de Fusion:–130 °C
Densité:0.63 g/cm³ (20 °C)
ONU:1265
:01-2119459286-30
:-40
:GHS02
:GHS08
:GHS07
:GHS09
Température d'entreposage:Température ambiante
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:99
Point d'éclair:–40 °C
:Boiling Pt:
:Boiling Pt: 35 - 36 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –130 °C
:Flash Pt:
:Flash Pt: -40 °C
:M.W. 72.15 g/mol
:3
:II
Température d'entreposage:Ambient
Customers Who Bought This Also Bought