Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH83626.400
Fournisseur: VWR International
Numéro CAS: 67-66-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:4 L
- Cat. no.:BDH83626.400
- Supplier No.:BDH83626.400
Spécifications
Résultats des tests de spécification
- Assay (by GC-FID):Min. 99.8 %
- Water (by KF coulometry):Max. 0.05 %
- Residue after evaporation:Max. 0.001 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (250 nm):Min. 50 %
- Transmittance (260 nm):Min. 84 %
- Transmittance (280 nm):Min. 95 %
- Transmittance (300 nm):Min. 98 %
- Conforms to BDH 15283:Passes Test
- Stabilizer (as Amylenes):100 - 200 ppm
- Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Methane, trichloro-,Trichloroform
Point d'ébullition:61,7 °C (1013 hPa)
Point de Fusion:-63 °C
Densité:1,4832 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:stabilisé avec 1-pentène
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 61,7 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -63 °C
:Pt Eclair:|Point eclaire:
:M.W. 119,38 g/mol
Index Merck:13,02160
Point de Fusion:–63 °C
Densité:1.4832 g/cm³ (20 °C)
EINECS:200-663-8
:-63
:61,7
Température d'entreposage:Température ambiante
ONU:1888
:05-2114614906-45
:GHS06
:GHS08
:Storage Temperature: Ambient
Point d'ébullition:61.7 °C (1013 hPa)
:99,8
:Stabilised with amylene
:Boiling Pt:
:Boiling Pt: 61.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –63 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.38 g/mol
:61.7
:6.1
:III
Température d'entreposage:Ambient
:Amylen
Customers Who Bought This Also Bought