Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
# de catalogue BDH20067.400
Fournisseur: VWR International
Numéro CAS: 67-64-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:4 L
- Supplier No.:BDH20067.400
Spécifications
Résultats des tests de spécification
- Assay (by GC-FID) (Corrected for Water):Min. 99.5 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Residue after evaporation:Max. 5 ppm
- Water (by KF coulometry):Max. 0.2 %
- Transmittance (345 nm):Min. 50 %
- Transmittance (350 nm):Min. 80 %
- Transmittance (360 nm):Min. 98 %
- VWR Chemicals BDH®, Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC:
Résultats des tests de spécification
A propos de cet article
Synonymes: 2-Propanone,DMK,Propanone
Formule:CH₃COCH₃
Point d'ébullition:56,2 °C (1013 hPa)
Point de Fusion:-95,4 °C
Densité:0,792 g/cm³ (20 °C)
Point d'éclair:<–20 °C (closed cup)
Numéro MDL:MFCD00008765
Numéro CAS:67-64-1
:Storage Temperature: Ambient
ADR:3,II
:Length
:Width
:Height
:Diameter
:Thickness
:99,5
:Point d'ébullition:
:Pt ébul.: 56,2 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -95,4 °C
:Point eclaire:
:Pt Eclair: Max. -20 °C (coupe fermée)
:M.W. 58,08 g/mol
Index Merck:13,00067
EINECS:200-662-2
:-95,4
:56,2
ONU:1090
:01-2119471330-49
:3
:II
:≤ -20 °C (closed cup)
:GHS02
:GHS07
Température d'entreposage:Température ambiante
Customers Who Bought This Also Bought