ContactUsLinkComponent
FETALGRO BOVINE GROWTH SERUM 500ML
Fetalgro Bovine Growth Serum, RMBIO
# de catalogue 10221-866
FETALGRO BOVINE GROWTH SERUM 500ML
Fetalgro Bovine Growth Serum, RMBIO
# de catalogue 10221-866
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Serum format:
    Frozen
  • Description:
    Fetalgro Bovine Growth Serum
  • Size:
    500 mL
  • Cat. no.:
    10221-866
  • Packaging:
    Nalgene bottle
  • Supplier No.:
    FGR-BBT
Spécifications
A propos de cet article
Fetalgro® is Rocky Mountain Biologicals proprietary bovine growth serum (BGS) supplemented with additives that match or outperform FBS' cell growth capabilities in many cell lines.
Each lot of Fetalgro® is tested by RMBIO's team of experienced scientists in our own state-of-the-art cell culture lab prior to releasing the product for customer use. This investment in Fetalgro's product release testing gives us the ability to find the optimal performance for specific cell lines.
Certifications: Tested according to 9CFR 113.26, 28, 53
Customers Who Bought This Also Bought