Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
# de catalogue 10218-470
Fournisseur: Thermo Fisher Scientific
Spécifications
- Ø:25 mm
- Pore size:0.20 µm
- Sterility:Sterile
- Cat. no.:10218-470
- Membrane:SFCA
- Supplier No.:723-2520
Spécifications
A propos de cet article
Filter sample volumes of 10 to 100 ml with these 25 mm Ø syringe filters.
- Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
- SFCA syringe filters have a blue housing ring
CA membranes have some protein binding and extractables, making them more suitable for clarification of buffers, aqueous solutions and reagents. CA syringe filters have a red housing ring. All housings are polypropylene with female Luer-Lok®inlet and male Luer slip outlet.
SFCA membranes are low protein binding and have lower extractables than standard CA so SFCA is a good choice for cell culture fluids and cellular solutions.
Packaging: Sterile filters are individually blister-packed. Non-sterile filters are bulk packed in zip closure bags.
Customers Who Bought This Also Bought