Order Entry
ContactUsLinkComponent
EDTA TEST STRIPS (100)
Quantofix EDTA Test Strips, CTL Scientific Supply
# de catalogue 10217-566
Fournisseur:  Macherey-Nagel
Numéro CAS:  
Quantofix EDTA Test Strips, CTL Scientific Supply
# de catalogue 10217-566
Fournisseur:  Macherey-Nagel
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    EDTA Test Strips
  • Storage:
    Room Temperature
  • Cat. no.:
    10217-566
  • Range:
    0-100-200-300-400 mg/L
  • Supplier No.:
    91335

Spécifications

A propos de cet article

Quantofix EDTA test strips is for the rapid and reliable determination of EDTA, NTA and other complexing agents in solutions

Quantofix EDTA has a gradation of scale: 0-100-200-300-400 mg/L EDTA.

The easy dip and read procedure provides a reliable result within 15 seconds.

Ordering information: Packaged as a box of 100.
Customers Who Bought This Also Bought