To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Length:25.4 mm (1.00")
- Description:SurGuard 3 Safety Needle
- Gauge:18
- Supplier No.:SG3-1825
Spécifications
A propos de cet article
SurGuard 3 safety needles offer 3 ways to activate and a broader range of sizes than most other brands
With separate thumb and finger grips, sharper needles, and competitive pricing, SurGuard 3 fits every clinician’s style.