Order Entry
ContactUsLinkComponent
Universal pH indicator paper, pH range 1-14, box of 200 strips 10mm by 70mm
pH Indicator Papers and Strips, Ahlstrom
# de catalogue 10199-618
Fournisseur:  Ahlstrom
Universal pH indicator paper, pH range 1-14, box of 200 strips 10mm by 70mm
pH Indicator Papers and Strips, Ahlstrom
# de catalogue 10199-618
Fournisseur:  Ahlstrom

Spécifications

  • Description:
    Universal pH Indicator Paper Strips, 10 mm x 70 mm
  • Cat. no.:
    10199-618
  • pH Range:
    pH 1 - 14
  • Supplier No.:
    PHB2-0114

Spécifications

A propos de cet article

Ahlstrom offers a range of test papers for rapid determination of pH

Universal pH Indicator papers are availabe in a roll of 5 m in a plastic case, a refill pack of 3 reels for that case, and a box of 200 strips. pH test strips, which have small squares of paper, each prepared with a different indicator for more accurate determination of pH are also available. All include color charts.

Customers Who Bought This Also Bought