ContactUsLinkComponent
Glass Microfiber Filter
Glass Microfiber Filter Paper, Grade 169, Ahlstrom
# de catalogue 10199-616
Fournisseur:  Ahlstrom
Glass Microfiber Filter
Glass Microfiber Filter Paper, Grade 169, Ahlstrom
# de catalogue 10199-616
Fournisseur:  Ahlstrom
Spécifications
  • Flow rate:
    Fast
  • Size:
    9.0 cm dia.
  • Retention:
    1.0 microns
  • Load capacity:
    Very High
  • Cat. no.:
    10199-616
  • Supplier No.:
    1690-0900
Spécifications
A propos de cet article
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 degrees Centrigrade, as required for testing for volatile solids in STM 2540.
Customers Who Bought This Also Bought