Glass Microfiber Filter Paper, Grade 169, Ahlstrom
# de catalogue 10199-616
Fournisseur: Ahlstrom
Spécifications
- Flow rate:Fast
- Size:9.0 cm dia.
- Retention:1.0 microns
- Load capacity:Very High
- Cat. no.:10199-616
- Supplier No.:1690-0900
Spécifications
A propos de cet article
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 degrees Centrigrade, as required for testing for volatile solids in STM 2540.
Customers Who Bought This Also Bought