Glass Extraction Thimbles, Ahlstrom
# de catalogue 10199-568
Fournisseur: Ahlstrom
Spécifications
- Length:100 mm
- Inner Diameter:30 mm
- Supplier No.:E160-3010
Spécifications
A propos de cet article
Glass thimbles are used to sample dust particles and aerosols from gaseous streams or for Soxhlet extractions requiring solvents too aggressive for cellulose thimbles
Wall thickness is approximately 2 mm thick. Temperature resistance of 500 degrees Celsius.
Customers Who Bought This Also Bought