Spécifications
- Material:Stainless Steel
- Cat. no.:10195-074
- Length:114 mm (4 1/2")
- Supplier No.:23-1104
- Blade Type:Straight, Sharp/Blunt
Spécifications
A propos de cet article
Sklarlite™ Operating Scissors, also known as Surgical Scissors are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying Blade Types and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.