Spécifications
- Sterility:Non sterile
- Description:Pederson Vaginal Speculum, Large, 4.75×1"
- Latex/Latex-free:Latex-free
- Material:Stainless Steel
- Curvature:Straight
- Grade:Premium OR-Grade
- Supplier No.:90-3711
Spécifications
A propos de cet article
Sklar® Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed.
- The blades are then opened by an external lever and screw
- Reusable and latex-free
The blades of the Pederson speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.