ContactUsLinkComponent
Speculum
Pederson Vaginal Speculum, OR Grade, Sklar
# de catalogue 10193-698
Fournisseur:  Sklar
Speculum
Pederson Vaginal Speculum, OR Grade, Sklar
# de catalogue 10193-698
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sterility:
    Non sterile
  • Description:
    Pederson Vaginal Speculum, Large, 4.75×1"
  • Latex/Latex-free:
    Latex-free
  • Material:
    Stainless Steel
  • Curvature:
    Straight
  • Grade:
    Premium OR-Grade
  • Supplier No.:
    90-3711
Spécifications
A propos de cet article
Sklar® Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed.
  • The blades are then opened by an external lever and screw
  • Reusable and latex-free
The blades of the Pederson speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.
Customers Who Bought This Also Bought