ContactUsLinkComponent
Forcep
Merit™ Lorna (Edna) Towel Clamps, Physician Grade, Sklar
# de catalogue 10184-878
Fournisseur:  Sklar
Forcep
Merit™ Lorna (Edna) Towel Clamps, Physician Grade, Sklar
# de catalogue 10184-878
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    89 mm (31/2")
  • Material:
    Stainless Steel
  • Cat. no.:
    10184-878
  • Supplier No.:
    97-629
Spécifications
A propos de cet article
Merit™ Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site.
The jaws and tips are curved while the shanks and handle are flat, so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly refered to as the Edna Towel Clamp. This clamp should never be used on skin.
Merit™ instruments are mid-grade products used for routine and in-office minor procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee.
Customers Who Bought This Also Bought