Spécifications
- Length:89 mm (31/2")
- Material:Stainless Steel
- Cat. no.:10184-878
- Supplier No.:97-629
Spécifications
A propos de cet article
Merit™ Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site.
The jaws and tips are curved while the shanks and handle are flat, so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly refered to as the Edna Towel Clamp. This clamp should never be used on skin.
Merit™ instruments are mid-grade products used for routine and in-office minor procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee.