Spécifications
- Description:Endothelial Cell Growth Medium MV 2 Kit
- Size:500 ml
- Cat. no.:10175-256
- Supplier No.:C-22121
Spécifications
A propos de cet article
PromoCell Endothelial Cell Growth Medium MV and Endothelial Cell Growth Medium MV2 were developed for the in vitro cultivation of endothelial cells from microvascular vessels, the coronary artery, and the aorta
The media are optimized for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.
The media are optimized for primary human cells but can also be used for bovine, porcine, murine, rat and dog endothelial cells from microvascular vessels as well as for various endothelial cell lines.
Recommended for Human Dermal Microvascular Endothelial Cells (HDMEC), Human Cardiac Microvascular Endothelial Cells (HCMEC), Human Coronary Artery Endothelial Cells (HCAEC), Human Aortic Endothelial Cells (HAoEC), Human Pulmonary Microvascular Endothelial Cells (HPMEC), Human Dermal Lymphatic Endothelial Cells (HDLEC), Human Dermal Blood Endothelial Cells (HDBEC).
Endothelial Cell Growth Medium MV2 lacks the Endothelial Cell Growth Supplement (ECGS, bovine hypothalamic extract) but contains the basic Fibroblast Growth Factor, the Insulin-like Growth Factor (Long R3 IGF-1) and the Vascular Endothelial Growth Factor (VEGF). Generally, VEGF leads to higher endothelial cell proliferation in culture. But, because of its multiple effects on the cell metabolism, it may also interfere with certain experimental setups. In these cases, we recommend the use of Endothelial Cell Growth Medium MV.