Spécifications
- Description:Airway Epithelial Cell Growth Medium (Ready-to-use)
- Size:500 ml
- Cat. no.:10175-240
- Supplier No.:C-21060
Spécifications
A propos de cet article
The PromoCell Airway Epithelial Cell Growth Medium is a serum-free medium optimized for the cultivation of epithelial cells from large air passages
Recommended for Human Nasal Epithelial Cells (HNEpC), Human Bronchial Epithelial Cells (HBEpC), Human Tracheal Epithelial Cells (HTEpC).
Recommended for Human Nasal Epithelial Cells (HNEpC), Human Bronchial Epithelial Cells (HBEpC), Human Tracheal Epithelial Cells (HTEpC).
The medium is optimized for primary human cells but can also be used for murine, porcine, equine, and rat airway epithelial cells.