ContactUsLinkComponent
Mesenchymal Stem Cell Osteogenic Different. Medium
Mesenchymal Stem Cell Medium, PromoCell
# de catalogue 10172-378
Fournisseur:  Promocell
Numéro CAS:  
Mesenchymal Stem Cell Osteogenic Different. Medium
Mesenchymal Stem Cell Medium, PromoCell
# de catalogue 10172-378
Fournisseur:  Promocell
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Mesenchymal Stem Cell Osteogenic Differentiation Medium (Ready-to-use)
  • Size:
    100 ml
  • Cat. no.:
    10172-378
  • Supplier No.:
    C-28013
Spécifications
A propos de cet article
PromoCell Mesenchymal Stem Cell Media were developed for the in vitro expansion and directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue
The Mesenchymal Stem Cell Growth Medium DXF allows for the robust proliferation of multipotent MSC in a chemically defined and xeno-free culture environment. It is devoid of all animal-derived components and substances of human origin with Human Serum Albumin as the only exception. Fibronectin-coated plates are necessary in conjunction with this medium. In combination with the Cell Dissociation Solution ACF (C-41320) and human fibronectin (C-43060), the PromoCell MSC Growth Medium DXF provides a completely xeno-free and defined culture system for human MSC.
  • Supports the expansion of multipotent human Mesenchymal Stem Cells (MSC)
  • In vitro expansion from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue
  • Five media to efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages
The PromoCell Mesenchymal Stem Cell Growth Medium supports the expansion of multipotent human Mesenchymal Stem Cells (MSC) without inducing early senescence and differentiation as observed in standard culture media. PromoCell offers five media to efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively. Note: The PromoCell MSC Chondrogenic Differentiation Medium w/o Inducers (C-28014) must be supplemented with adequate chondrogenic inducers by the customer. Recommended for Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM), Human Mesenchymal Stem Cells from Umbilical Cord Matrix (hMSC-UC), Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT).
Ordering information: PromoCell Mesenchymal Stem Cell Media are available as Medium (Readyto- use) and consist of a bottle of Basal Medium and one vial of Supplement- Mix. Adding the SupplementMix to the Basal Medium results in the complete Medium.
Customers Who Bought This Also Bought