Order Entry
ContactUsLinkComponent
Fibroblast Basal Medium 2: Fibroblast Basal Medium 2; 500 ml
Fibroblast Growth Medium 2 for Adult Cells, PromoCell
# de catalogue 10172-066
Fournisseur:  Promocell
Numéro CAS:  
Fibroblast Basal Medium 2: Fibroblast Basal Medium 2; 500 ml
Fibroblast Growth Medium 2 for Adult Cells, PromoCell
# de catalogue 10172-066
Fournisseur:  Promocell
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Fibroblast Basal Medium 2
  • Size:
    500 ml
  • Cat. no.:
    10172-066
  • Supplier No.:
    C-23220

Spécifications

A propos de cet article

PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium developed for adult human fibroblasts

Recommended for Normal Human Dermal Fibroblasts (NHDF), adult donor, Human Pulmonary Fibroblasts (HPF), Human Aortic Adventitial Fibroblasts (HAoAF), Human Uterine Fibroblasts (HUF), Human Villious Mesenchymal Fibroblasts (HVMF)

Recommended for Normal Human Dermal Fibroblasts (NHDF), adult donor, Human Pulmonary Fibroblasts (HPF), Human Aortic Adventitial Fibroblasts (HAoAF), Human Uterine Fibroblasts (HUF), Human Villious Mesenchymal Fibroblasts (HVMF)

The medium is optimized for primary human cells but can also be used for rat, porcine, and bovine fibroblasts as well as fibroblast cell lines

Ordering information: PromoCell Fibroblast Growth Medium 2 is available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the Supplement- Pack (a set of individual vials with premeasured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.
Customers Who Bought This Also Bought