Order Entry
ContactUsLinkComponent
Skeletal Muscle Cell Growth Medium KIT: Skeletal Muscle Cell Growth Medium Kit; 500 ml
Skeletal Muscle Cell Growth Medium, PromoCell
Catalog # 10172-060
Supplier:  Promocell
CAS Number:  
Skeletal Muscle Cell Growth Medium KIT: Skeletal Muscle Cell Growth Medium Kit; 500 ml
Skeletal Muscle Cell Growth Medium, PromoCell
Catalog # 10172-060
Supplier:  Promocell
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Skeletal Muscle Cell Growth Medium Kit
  • Size:
    500 ml
  • Cat. no.:
    10172-060
  • Supplier No.:
    C-23160

Specifications

About this item

The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum (5% V/V) medium optimized for the expansion of human skeletal muscle cells

The unique Skeletal Muscle Cell Differentiation Medium is a serum-free medium that induces the fusion of myoblasts to myotubes with typical multinucleated syncytia. The media are optimized for primary human cells but can also be used for rat, murine, and rabbit skeletal muscle cells.

Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the Supplements to the Basal Medium results in the complete Growth or Differentiation Medium, respectively. Basal Medium (with or without phenol red) as well as SupplementMix (Growth and Differentiation Medium) and SupplementPack (Growth Medium) can also be purchased separately.
Customers Who Bought This Also Bought