Spécifications
- Description:Skeletal Muscle Cell Differentiation Medium (Ready-to-use)
- Size:500 ml
- Cat. no.:10172-052
- Supplier No.:C-23061
Spécifications
A propos de cet article
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum (5% V/V) medium optimized for the expansion of human skeletal muscle cells
The unique Skeletal Muscle Cell Differentiation Medium is a serum-free medium that induces the fusion of myoblasts to myotubes with typical multinucleated syncytia. The media are optimized for primary human cells but can also be used for rat, murine, and rabbit skeletal muscle cells.